Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017994.4 | NP_060464.1 | 2, 3 | 419 | ||
XM_005250482.3 | XP_005250539.1 | 2, 3 | 271 | |||
GenBank mRNA | AK000961.1 | - | 2, 3 | 405 | ||
AK222774.1 | - | 2, 3 | 419 | |||
BC008675.1 | - | 2, 3 | 399 | 3853834 | ||
BC010519.1 | - | 2, 3 | 373 | 4150551 | ||
BC012562.1 | - | 2, 3 | 414 | 4332094 | ||
BC041764.2 | - | 2, 3 | 388 | 4152659 |