Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310332.1 | NP_001297261.1 | 3 | 198 | ||
NM_017999.4 | NP_060469.4 | 3 | 606 | |||
GenBank mRNA | AB265810.1 | - | 3 | 357 | ||
AK122711.1 | - | 2 | 59 | |||
AK225832.1 | - | 3 | 556 | |||
AK291247.1 | - | 3 | 558 | |||
AY256461.1 | - | 3 | 186 | |||
BC012077.1 | - | 3 | 526 | 4653017 | ||
DA462121.1 | - | 3 | 198 |