Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321829.1 | NP_001308758.1 | 3 | 820 | ||
NM_018017.3 | NP_060487.2 | 2 | 695 | |||
NM_153249.1 | NP_694981.1 | 3 | 820 | |||
NR_135815.1 | 3 | 820 | ||||
XM_011539916.2 | XP_011538218.1 | 2 | 787 | |||
GenBank mRNA | AF273054.1 | - | 1 | 210 | ||
AK092620.1 | - | 3 | 803 | |||
AK307517.1 | - | 2 | 672 | |||
BC020842.1 | - | 2 | 687 | |||
BC070368.1 | - | 2 | 666 | |||
BC100670.1 | - | 3 | 747 | |||
BC103499.1 | - | 2 | 622 | 40001826 | ||
BC105632.1 | - | 2 | 622 | 40001824 | ||
BC107430.1 | - | 2 | 655 | |||
BC152306.1 | - | 2 | 626 | 40012131 | ||
BX537964.1 | - | 2 | 667 |