Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018043.5 | NP_060513.5 | 16 | 1850 | ||
NR_030691.1 | 14 | 1608 | ||||
XM_006718602.2 | XP_006718665.1 | 17 | 1632 | |||
XM_006718604.2 | XP_006718667.1 | 17 | 1747 | |||
XM_006718605.2 | XP_006718668.1 | 14 | 1603 | |||
XM_011545121.2 | XP_011543423.1 | 17 | 1624 | |||
XM_011545123.2 | XP_011543425.1 | 17 | 1621 | |||
XM_011545124.2 | XP_011543426.1 | 16 | 1612 | |||
XM_011545125.2 | XP_011543427.1 | 16 | 1546 | |||
XM_011545126.2 | XP_011543428.1 | 15 | 1534 | |||
XM_011545127.2 | XP_011543429.1 | 15 | 1480 | |||
XM_011545128.2 | XP_011543430.1 | 14 | 1468 | |||
XM_011545129.2 | XP_011543431.1 | 17 | 1539 | |||
XM_011545131.2 | XP_011543433.1 | 14 | 1476 | |||
XM_017017956.1 | XP_016873445.1 | 16 | 1555 | |||
XM_017017957.1 | XP_016873446.1 | 15 | 1546 | |||
GenBank mRNA | AB845669.1 | - | 16 | 1545 | ||
AK001123.1 | - | 6 | 388 | |||
AK097619.1 | - | 14 | 1476 | |||
AK293088.1 | - | 16 | 1558 | |||
AL833582.1 | - | 16 | 1594 | |||
AY728143.1 | - | 16 | 1850 | |||
BC027590.2 | - | 10 | 724 | |||
BC033036.2 | - | 14 | 1564 | 4823645 |