Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143888.2 | NP_001137360.1 | 10 | 1326 | ||
NM_001143889.2 | NP_001137361.1 | 9 | 1107 | |||
NM_001143890.2 | NP_001137362.1 | 8 | 990 | |||
NM_001300958.1 | NP_001287887.1 | 10 | 1326 | |||
NM_018045.7 | NP_060515.3 | 10 | 1275 | |||
NR_125352.1 | 11 | 1384 | ||||
XM_005270986.4 | XP_005271043.1 | 10 | 1255 | |||
XM_011541677.2 | XP_011539979.1 | 10 | 1573 | |||
GenBank mRNA | AK001138.1 | - | 10 | 1245 | ||
AK225026.1 | - | 10 | 1256 | |||
AK225717.1 | - | 10 | 1245 | |||
AK297352.1 | - | 9 | 1071 | |||
AK297604.1 | - | 9 | 1076 | |||
AK300231.1 | - | 10 | 1184 | |||
AK300306.1 | - | 10 | 1295 | |||
AK304477.1 | - | 8 | 970 | |||
AK316172.1 | - | 9 | 1088 | |||
AY358230.1 | - | 1 | 277 | |||
BC010573.1 | - | 10 | 1234 | |||
BC026322.1 | - | 10 | 1351 | 4794111 | ||
BC028381.2 | - | 10 | 1247 | 4812029 | ||
BM760887.1 | - | 1 | 36 | |||
BX641056.1 | - | 9 | 1292 | |||
CR457252.1 | - | 9 | 1228 | |||
CR749228.1 | - | 9 | 1178 |