Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143888.2 | NP_001137360.1 | 2 | 99 | ||
NM_001143889.2 | NP_001137361.1 | 2 | 99 | |||
NM_001143890.2 | NP_001137362.1 | 2 | 99 | |||
NM_001300958.1 | NP_001287887.1 | 2 | 99 | |||
NM_018045.7 | NP_060515.3 | 2 | 99 | |||
NR_125352.1 | 2 | 99 | ||||
XM_005270986.4 | XP_005271043.1 | 2 | 79 | |||
XM_011541677.2 | XP_011539979.1 | 2 | 69 | |||
GenBank mRNA | AK001138.1 | - | 2 | 69 | ||
AK021440.1 | - | 2 | 78 | |||
AK225026.1 | - | 2 | 80 | |||
AK225717.1 | - | 2 | 69 | |||
AK293454.1 | - | 2 | 69 | |||
AK297352.1 | - | 2 | 78 | |||
AK297604.1 | - | 2 | 68 | |||
AK300231.1 | - | 2 | 66 | |||
AK300306.1 | - | 2 | 68 | |||
AK304477.1 | - | 2 | 79 | |||
AK316172.1 | - | 2 | 80 | |||
BC010573.1 | - | 2 | 58 | |||
BC026322.1 | - | 1 | 66 | 4794111 | ||
BC028381.2 | - | 2 | 71 | 4812029 | ||
BP363955.1 | - | 2 | 99 | |||
BX641056.1 | - | 1 | 65 | |||
CR457252.1 | - | 1 | 52 | |||
CR749228.1 | - | 1 | 60 |