Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143888.2 | NP_001137360.1 | 7 | 653 | ||
NM_001143889.2 | NP_001137361.1 | 6 | 434 | |||
NM_001143890.2 | NP_001137362.1 | 5 | 317 | |||
NM_001300958.1 | NP_001287887.1 | 7 | 653 | |||
NM_018045.7 | NP_060515.3 | 7 | 602 | |||
NR_125352.1 | 7 | 602 | ||||
XM_005270986.4 | XP_005271043.1 | 7 | 582 | |||
XM_011541677.2 | XP_011539979.1 | 7 | 900 | |||
GenBank mRNA | AK001138.1 | - | 7 | 572 | ||
AK225026.1 | - | 7 | 583 | |||
AK225717.1 | - | 7 | 572 | |||
AK293454.1 | - | 6 | 367 | |||
AK297604.1 | - | 6 | 403 | |||
AK300231.1 | - | 7 | 511 | |||
AK300306.1 | - | 7 | 622 | |||
AK304477.1 | - | 5 | 297 | |||
AK316172.1 | - | 6 | 415 | |||
BC010573.1 | - | 7 | 561 | |||
BC026322.1 | - | 6 | 569 | 4794111 | ||
BC028381.2 | - | 7 | 574 | 4812029 | ||
BX641056.1 | - | 6 | 619 | |||
CR457252.1 | - | 6 | 555 | |||
CR749228.1 | - | 6 | 505 |