Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018051.4 | NP_060521.4 | 5 | 980 | ||
XM_005249549.1 | XP_005249606.1 | 3 | 619 | |||
XM_006716041.2 | XP_006716104.1 | 5 | 995 | |||
XM_011516367.1 | XP_011514669.1 | 5 | 837 | |||
XM_011516368.2 | XP_011514670.1 | 5 | 995 | |||
XM_011516369.2 | XP_011514671.1 | 5 | 826 | |||
XM_017012381.1 | XP_016867870.1 | 5 | 1017 | |||
XM_017012382.1 | XP_016867871.1 | 5 | 995 | |||
XM_017012383.1 | XP_016867872.1 | 5 | 1056 | |||
XM_017012384.1 | XP_016867873.1 | 3 | 619 | |||
XM_017012385.1 | XP_016867874.1 | 3 | 619 | |||
XM_017012386.1 | XP_016867875.1 | 3 | 619 | |||
XM_017012387.1 | XP_016867876.1 | 3 | 619 | |||
GenBank mRNA | BC014491.1 | - | 5 | 980 | 4906683 | |
DB072968.1 | - | 3 | 456 |