Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114636.1 | NP_001108108.1 | 13 | 1122 | ||
NM_018062.3 | NP_060532.2 | 13 | 1107 | |||
XM_005264395.3 | XP_005264452.1 | 13 | 1107 | |||
XM_005264397.4 | XP_005264454.1 | 9 | 826 | |||
XM_011532939.2 | XP_011531241.1 | 14 | 1167 | |||
XM_011532940.2 | XP_011531242.1 | 14 | 1152 | |||
XM_011532941.1 | XP_011531243.1 | 14 | 1167 | |||
XM_011532942.2 | XP_011531244.1 | 13 | 1122 | |||
XM_011532944.2 | XP_011531246.1 | 10 | 844 | |||
XM_011532945.2 | XP_011531247.1 | 9 | 812 | |||
XM_017004414.1 | XP_016859903.1 | 14 | 1152 | |||
XM_017004416.1 | XP_016859905.1 | 9 | 826 | |||
GenBank mRNA | AK001197.1 | - | 13 | 1073 | ||
AK225147.1 | - | 12 | 963 | |||
AK309671.1 | - | 10 | 922 | |||
BC009042.1 | - | 13 | 1077 | 3870828 | ||
BC054517.1 | - | 13 | 1082 | 5180184 | ||
BQ574618.1 | - | 2 | 622 |
Gene Symbol: VRK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005264540.4 | XP_005264597.1 | 13 | 2235 | ||
XM_006712090.3 | XP_006712153.1 | 14 | 2074 | |||
XM_006712091.3 | XP_006712154.1 | 14 | 2319 | |||
XM_006712092.3 | XP_006712155.1 | 14 | 2407 | |||
XM_006712093.3 | XP_006712156.1 | 11 | 2112 | |||
XM_011533092.2 | XP_011531394.1 | 13 | 2019 | |||
XM_017004858.1 | XP_016860347.1 | 14 | 2219 | |||
XM_017004859.1 | XP_016860348.1 | 14 | 2897 | |||
XM_017004860.1 | XP_016860349.1 | 13 | 2827 | |||
XM_017004861.1 | XP_016860350.1 | 11 | 1926 | |||
XM_017004862.1 | XP_016860351.1 | 12 | 2739 | |||
XM_017004863.1 | XP_016860352.1 | 13 | 2319 | |||
XM_017004864.1 | XP_016860353.1 | 15 | 2451 | |||
XM_017004865.1 | XP_016860354.1 | 15 | 2270 | |||
XM_017004866.1 | XP_016860355.1 | 15 | 2319 |