Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114636.1 | NP_001108108.1 | 8, 9 | 765 | ||
NM_018062.3 | NP_060532.2 | 8, 9 | 750 | |||
XM_005264395.3 | XP_005264452.1 | 8, 9 | 750 | |||
XM_005264397.4 | XP_005264454.1 | 4, 5 | 469 | |||
XM_011532939.2 | XP_011531241.1 | 9, 10 | 810 | |||
XM_011532940.2 | XP_011531242.1 | 9, 10 | 795 | |||
XM_011532941.1 | XP_011531243.1 | 9, 10 | 810 | |||
XM_011532942.2 | XP_011531244.1 | 8, 9 | 765 | |||
XM_011532944.2 | XP_011531246.1 | 5, 6 | 487 | |||
XM_011532945.2 | XP_011531247.1 | 4, 5 | 455 | |||
XM_017004414.1 | XP_016859903.1 | 9, 10 | 795 | |||
XM_017004416.1 | XP_016859905.1 | 4, 5 | 469 | |||
GenBank mRNA | AK001197.1 | - | 8, 9 | 716 | ||
AK297736.1 | - | 6, 7 | 559 | |||
AK309671.1 | - | 5, 6 | 565 | |||
BC009042.1 | - | 8, 9 | 720 | 3870828 | ||
BC054517.1 | - | 8, 9 | 725 | 5180184 |