Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032142.3 | NP_115518.3 | 17, 18 | 3106 | ||
XM_005258107.3 | XP_005258164.1 | 17, 18 | 3161 | |||
XM_005258109.3 | XP_005258166.1 | 16, 17 | 2951 | |||
XM_005258110.1 | XP_005258167.1 | 4, 5 | 1345 | |||
XM_006722326.3 | XP_006722389.1 | 17, 18 | 3161 | |||
XM_006722327.3 | XP_006722390.1 | 16, 17 | 2951 | |||
XM_006722330.3 | XP_006722393.1 | 11, 12 | 2144 | |||
XM_011525673.2 | XP_011523975.1 | 17, 18 | 3161 | |||
XM_011525675.2 | XP_011523977.1 | 14, 15 | 2710 | |||
XM_017025803.1 | XP_016881292.1 | 16, 17 | 2996 | |||
XM_017025804.1 | XP_016881293.1 | 11, 12 | 2144 | |||
XM_017025805.1 | XP_016881294.1 | 6, 7 | 1485 | |||
XM_017025806.1 | XP_016881295.1 | 14, 15 | 2591 | |||
XM_017025807.1 | XP_016881296.1 | 10, 11 | 2203 | |||
GenBank mRNA | AK074074.1 | - | 2, 3 | 610 | ||
AK096548.1 | - | 14, 15 | 2329 | |||
AL136818.1 | - | 2, 3 | 759 | |||
BC144480.1 | - | 7, 8 | 1642 | 9053007 | ||
BC144481.1 | - | 8, 9 | 1852 | 9053008 | ||
BX648232.1 | - | 12, 13 | 2417 | |||
BX649151.1 | - | 4, 5 | 1345 | |||
KJ567064.1 | - | 16, 17 | 3026 |