Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304496.1 | NP_001291425.1 | 5 | 694 | ||
NM_018073.7 | NP_060543.5 | 6 | 1177 | |||
GenBank mRNA | AF360739.1 | - | 6 | 1130 | ||
AF439153.1 | - | 5 | 887 | |||
AK001231.1 | - | 3 | 382 | |||
AK022923.1 | - | 3 | 347 | |||
AK056523.1 | - | 6 | 1139 | |||
AK291166.1 | - | 6 | 1089 | |||
AK301120.1 | - | 4 | 409 | |||
AL832730.1 | - | 6 | 1127 | |||
AY005802.1 | - | 5 | 563 | |||
BC075058.2 | - | 5 | 911 | 30915367 | ||
BC109063.2 | - | 6 | 1096 | 40033795 | ||
DA833140.1 | - | 5 | 577 |