Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166135.1 | NP_001159607.1 | 15 | 1919 | ||
NM_018077.2 | NP_060547.2 | 19 | 2342 | |||
XM_011516370.2 | XP_011514672.1 | 20 | 2449 | |||
XM_017012389.1 | XP_016867878.1 | 18 | 2225 | |||
XM_017012390.1 | XP_016867879.1 | 17 | 2117 | |||
GenBank mRNA | AK001239.1 | - | 19 | 2342 | ||
AK222716.1 | - | 19 | 2304 | |||
AK300500.1 | - | 15 | 1881 | |||
BC013889.2 | - | 19 | 2296 | 3926178 | ||
BU609187.1 | - | 1 | 478 | |||
KU178685.1 | - | 19 | 2227 | |||
KU178686.1 | - | 4 | 429 |