Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284220.1 | NP_001271149.1 | 9, 10 | 1157 | ||
NM_001284222.1 | NP_001271151.1 | 10, 11 | 1192 | |||
NM_001284224.1 | NP_001271153.1 | 8, 9 | 1054 | |||
NM_001284225.1 | NP_001271154.1 | 8, 9 | 1093 | |||
NM_001284226.1 | NP_001271155.1 | 9, 10 | 1118 | |||
NM_018091.5 | NP_060561.3 | 10, 11 | 1257 | |||
XM_006716354.2 | XP_006716417.1 | 9, 10 | 1147 | |||
XM_017013604.1 | XP_016869093.1 | 10, 11 | 1257 | |||
GenBank mRNA | AK001284.1 | - | 10, 11 | 1184 | ||
AK022626.1 | - | 10, 11 | 1213 | |||
AK223047.1 | - | 9, 10 | 1043 | |||
AK293424.1 | - | 8, 9 | 1020 | |||
AK295574.1 | - | 9, 10 | 1046 | |||
AK296475.1 | - | 9, 10 | 1112 | |||
AK298265.1 | - | 9, 10 | 1008 | |||
AK302140.1 | - | 8, 9 | 1019 | |||
AK315985.1 | - | 8, 9 | 997 | |||
AK316053.1 | - | 9, 10 | 1061 | |||
AL834273.1 | - | 10, 11 | 1115 | |||
BC001240.1 | - | 10, 11 | 1147 | 3457039 | ||
BX648011.1 | - | 10, 11 | 1191 |