Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284220.1 | NP_001271149.1 | 9 | 992 | ||
NM_001284222.1 | NP_001271151.1 | 10 | 1027 | |||
NM_001284224.1 | NP_001271153.1 | 8 | 889 | |||
NM_001284225.1 | NP_001271154.1 | 8 | 928 | |||
NM_001284226.1 | NP_001271155.1 | 9 | 953 | |||
NM_018091.5 | NP_060561.3 | 10 | 1092 | |||
XM_006716354.2 | XP_006716417.1 | 9 | 982 | |||
XM_017013604.1 | XP_016869093.1 | 10 | 1092 | |||
GenBank mRNA | AK001284.1 | - | 10 | 1019 | ||
AK022626.1 | - | 10 | 1048 | |||
AK223047.1 | - | 9 | 878 | |||
AK293424.1 | - | 8 | 855 | |||
AK295574.1 | - | 9 | 881 | |||
AK296475.1 | - | 9 | 947 | |||
AK298265.1 | - | 9 | 843 | |||
AK302140.1 | - | 8 | 854 | |||
AK315985.1 | - | 8 | 832 | |||
AK316053.1 | - | 9 | 896 | |||
AL834273.1 | - | 10 | 950 | |||
BC001240.1 | - | 10 | 982 | 3457039 | ||
BX648011.1 | - | 10 | 1026 |