Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130447.1 | NP_001123919.1 | 5 | 496 | ||
NM_001323630.1 | NP_001310559.1 | 5 | 456 | |||
NM_001323631.1 | NP_001310560.1 | 7 | 678 | |||
NM_001323632.1 | NP_001310561.1 | 5 | 519 | |||
NM_018097.2 | NP_060567.1 | 6 | 589 | |||
XM_017022396.1 | XP_016877885.1 | 6 | 593 | |||
XM_017022397.1 | XP_016877886.1 | 6 | 560 | |||
GenBank mRNA | AK001322.1 | - | 6 | 547 | ||
AK022939.1 | - | 6 | 560 | |||
BC010903.1 | - | 6 | 568 | 4285311 | ||
DA923763.1 | - | 5 | 453 |