Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC8D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134479.1 | NP_001127951.1 | 3 | 2057 | ||
NM_018103.4 | NP_060573.2 | 3 | 2044 | |||
XM_011541689.1 | XP_011539991.1 | 4 | 2128 | |||
XM_017001599.1 | XP_016857088.1 | 3 | 2399 | |||
XM_017001600.1 | XP_016857089.1 | 3 | 1854 | |||
XM_017001601.1 | XP_016857090.1 | 3 | 2745 | |||
XM_017001602.1 | XP_016857091.1 | 3 | 2098 | |||
GenBank mRNA | AK001332.1 | - | 1 | 1110 | ||
AK092238.1 | - | 3 | 2020 | |||
AK222787.1 | - | 1 | 1071 | |||
AL831880.1 | - | 1 | 1516 | |||
AL832952.1 | - | 1 | 1475 | |||
AY358874.1 | - | 1 | 632 | |||
BC024159.2 | - | 3 | 2011 | 3958069 |