Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018129.3 | NP_060599.1 | 2 | 364 | ||
XM_005257500.3 | XP_005257557.1 | 2 | 554 | |||
XM_017024813.1 | XP_016880302.1 | 2 | 152 | |||
GenBank mRNA | AF468030.1 | - | 2 | 211 | ||
AK001397.1 | - | 2 | 316 | |||
AK223242.1 | - | 2 | 295 | |||
AK303665.1 | - | 2 | 316 | |||
AK303792.1 | - | 2 | 317 | |||
BC006525.2 | - | 2 | 251 | 2966366 | ||
DA186382.1 | - | 2 | 336 | |||
DB228906.1 | - | 2 | 365 | |||
FJ224333.1 | - | 2 | 211 |