Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145417.1 | NP_001138889.1 | 2 | 903 | ||
NM_018133.3 | NP_060603.2 | 2 | 1704 | |||
XM_005247571.3 | XP_005247628.1 | 2 | 1438 | |||
XM_006713684.3 | XP_006713747.1 | 2 | 921 | |||
XM_011512949.2 | XP_011511251.1 | 2 | 1008 | |||
GenBank mRNA | AB046805.1 | - | 2 | 1052 | ||
AK091956.1 | - | 2 | 1704 | |||
AK292557.1 | - | 2 | 1199 | |||
AK302491.1 | - | 2 | 870 | |||
AL834289.1 | - | 2 | 1022 | |||
BC032719.1 | - | 1 | 493 | |||
BC093764.1 | - | 2 | 1004 | 7939609 | ||
BC093790.1 | - | 2 | 1003 | 7939635 |