Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193343.1 | NP_001180272.1 | 3 | 213 | ||
NM_018135.3 | NP_060605.1 | 3 | 213 | |||
XM_006715134.2 | XP_006715197.1 | 3 | 189 | |||
GenBank mRNA | AB049952.1 | - | 3 | 190 | ||
AK001410.1 | - | 3 | 219 | |||
AK300757.1 | - | 3 | 184 | |||
AK307183.1 | - | 3 | 191 | |||
BC010364.1 | - | 3 | 188 | 4248325 | ||
DQ884400.1 | - | 3 | 180 |
Gene Symbol: ZNF589 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017006577.1 | XP_016862066.1 | 1 | 1148 | ||
GenBank mRNA | AF114816.1 | - | 3 | 168 | ||
AF114817.1 | - | 3 | 168 |