Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIC8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018157.2 | NP_060627.2 | 3 | 414 | ||
XM_005268998.3 | XP_005269055.1 | 3 | 440 | |||
XM_011538526.2 | XP_011536828.1 | 2 | 796 | |||
XM_011538527.2 | XP_011536829.1 | 2 | 702 | |||
XM_011538528.2 | XP_011536830.1 | 2 | 798 | |||
XM_011538529.2 | XP_011536831.1 | 2 | 793 | |||
XM_017019548.1 | XP_016875037.1 | 3 | 440 | |||
XM_017019549.1 | XP_016875038.1 | 2 | 305 | |||
GenBank mRNA | AK001482.1 | - | 3 | 321 | ||
AY221116.1 | - | 3 | 223 | |||
BC034220.1 | - | 4 | 457 | 5264088 | ||
BC132689.1 | - | 3 | 395 | 40146711 | ||
BC144056.1 | - | 3 | 395 | 9052576 | ||
DA325821.1 | - | 3 | 414 |