Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP1S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308363.1 | NP_001295292.1 | 2 | 323 | ||
NM_018174.5 | NP_060644.4 | 2 | 217 | |||
XM_017026930.1 | XP_016882419.1 | 2 | 317 | |||
XM_017026931.1 | XP_016882420.1 | 2 | 558 | |||
GenBank mRNA | AJ440784.1 | - | 2 | 169 | ||
AK027623.1 | - | 2 | 169 | |||
AK292555.1 | - | 2 | 187 | |||
AK294936.1 | - | 2 | 194 | |||
AL834233.1 | - | 2 | 172 | |||
BC067115.1 | - | 2 | 172 | |||
BC080547.1 | - | 2 | 171 | 5925583 | ||
BC113952.1 | - | 2 | 169 | 40027183 | ||
BG771546.1 | - | 2 | 180 | |||
DA392500.1 | - | 2 | 216 | |||
DB081409.1 | - | 2 | 217 | |||
DB097723.1 | - | 2 | 439 | |||
DC401920.1 | - | 2 | 323 | |||
DQ387861.1 | - | 2 | 175 |