Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018176.3 | NP_060646.2 | 8 | 1231 | ||
GenBank mRNA | AB067503.1 | - | 8 | 1124 | ||
AF467955.1 | - | 8 | 1132 | |||
AJ487516.1 | - | 8 | 1132 | |||
AJ487958.1 | - | 8 | 1154 | |||
AK001537.1 | - | 1 | 174 | |||
AL833647.1 | - | 7 | 922 | |||
BC101759.1 | - | 8 | 1231 | 8069265 | ||
BC101761.1 | - | 8 | 1231 | 8069267 |