Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBNO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167856.1 | NP_001161328.1 | 7 | 998 | ||
NM_018183.3 | NP_060653.3 | 7 | 995 | |||
XM_005253572.3 | XP_005253629.1 | 8 | 1136 | |||
XM_005253573.4 | XP_005253630.1 | 9 | 1546 | |||
XM_005253575.3 | XP_005253632.1 | 8 | 1133 | |||
XM_005253576.3 | XP_005253633.1 | 9 | 1177 | |||
XM_006719473.3 | XP_006719536.1 | 10 | 1302 | |||
XM_006719474.3 | XP_006719537.1 | 9 | 1255 | |||
XM_011538533.2 | XP_011536835.1 | 10 | 1296 | |||
XM_011538534.2 | XP_011536836.1 | 7 | 1034 | |||
XM_011538535.2 | XP_011536837.1 | 7 | 1036 | |||
XM_017019556.1 | XP_016875045.1 | 8 | 1120 | |||
XM_017019557.1 | XP_016875046.1 | 8 | 1127 | |||
XM_017019558.1 | XP_016875047.1 | 9 | 1190 | |||
XM_017019559.1 | XP_016875048.1 | 8 | 1257 | |||
GenBank mRNA | AB014772.1 | - | 7 | 995 | ||
AK001695.1 | - | 8 | 1133 | |||
AK122703.1 | - | 9 | 1551 | |||
AK307488.1 | - | 7 | 1034 | |||
AY364255.1 | - | 7 | 997 | |||
BC030544.1 | - | 8 | 1129 | |||
BC133704.1 | - | 7 | 998 | 40124902 |