Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082969.1 | NP_001076438.1 | 5 | 458 | ||
NM_001082970.1 | NP_001076439.1 | 5 | 1016 | |||
NM_001301017.1 | NP_001287946.1 | 5 | 458 | |||
NM_001301019.1 | NP_001287948.1 | 5 | 499 | |||
NM_001301021.1 | NP_001287950.1 | 5 | 499 | |||
NM_018195.3 | NP_060665.3 | 5 | 1016 | |||
NR_103469.1 | 5 | 1057 | ||||
GenBank mRNA | AB096251.1 | - | 5 | 362 | ||
AK001588.1 | - | 2 | 74 | |||
AK094203.1 | - | 4 | 1816 | |||
AK125340.1 | - | 5 | 972 | |||
BC005403.1 | - | 3 | 255 | |||
BC030546.1 | - | 3 | 260 | |||
BC048133.1 | - | 5 | 944 | 5295844 | ||
BC072455.1 | - | 5 | 383 | 6185048 | ||
BG115155.1 | - | 5 | 506 | |||
BX538107.1 | - | 5 | 1015 | |||
CD244552.1 | - | 5 | 417 | |||
DA813095.1 | - | 5 | 444 | |||
EL736728.1 | - | 3 | 248 |