Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018218.2 | NP_060688.1 | 15 | 2087 | ||
XM_006712612.3 | XP_006712675.1 | 15 | 2105 | |||
XM_011511396.2 | XP_011509698.1 | 15 | 2105 | |||
XM_011511397.2 | XP_011509699.1 | 16 | 2179 | |||
XM_011511398.2 | XP_011509700.1 | 15 | 2105 | |||
XM_011511399.2 | XP_011509701.1 | 14 | 1906 | |||
XM_011511400.2 | XP_011509702.1 | 14 | 1890 | |||
XM_011511401.2 | XP_011509703.1 | 15 | 2105 | |||
XM_017004426.1 | XP_016859915.1 | 16 | 2179 | |||
XM_017004427.1 | XP_016859916.1 | 9 | 1107 | |||
XM_017004428.1 | XP_016859917.1 | 15 | 2147 | |||
GenBank mRNA | AJ583821.2 | - | 15 | 2087 | ||
AK001647.1 | - | 15 | 2049 | |||
AK297821.1 | - | 9 | 1175 |