Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC47A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018242.2 | NP_060712.2 | 3 | 349 | ||
GenBank mRNA | AK001709.1 | - | 3 | 349 | ||
AK222625.1 | - | 3 | 353 | |||
AK225360.1 | - | 3 | 327 | |||
AK302620.1 | - | 3 | 349 | |||
AK310270.1 | - | 3 | 168 | |||
BC010661.2 | - | 3 | 338 | 3874616 | ||
BC050592.1 | - | 3 | 323 | |||
BC058882.1 | - | 3 | 325 | 5188651 |