Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304530.1 | NP_001291459.1 | 5 | 343 | ||
NM_001304532.1 | NP_001291461.1 | 5 | 343 | |||
NM_018246.2 | NP_060716.2 | 6 | 395 | |||
XM_011544570.2 | XP_011542872.1 | 5 | 342 | |||
XM_011544571.2 | XP_011542873.1 | 4 | 221 | |||
GenBank mRNA | AK001715.1 | - | 5 | 343 | ||
AK027900.1 | - | 6 | 347 | |||
AK292875.1 | - | 5 | 330 | |||
BC006239.2 | - | 7 | 570 | |||
BC032588.1 | - | 5 | 332 | 5502227 | ||
BC049197.1 | - | 6 | 349 | 5534448 | ||
BC054039.1 | - | 6 | 335 | |||
BC065295.1 | - | 6 | 336 | 6139413 | ||
BG718771.1 | - | 5 | 335 | |||
DA218015.1 | - | 6 | 395 |