Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018266.2 | NP_060736.1 | 5 | 934 | ||
NR_073506.1 | 6 | 1043 | ||||
XM_005247578.1 | XP_005247635.1 | 5 | 899 | |||
XM_006713687.1 | XP_006713750.1 | 5 | 842 | |||
XM_011512958.2 | XP_011511260.1 | 5 | 899 | |||
XM_017006788.1 | XP_016862277.1 | 4 | 815 | |||
GenBank mRNA | AK001764.1 | - | 5 | 876 | ||
AK222950.1 | - | 5 | 866 | |||
AK223248.1 | - | 5 | 866 | |||
BC003192.1 | - | 6 | 958 | |||
BC010070.2 | - | 6 | 958 | |||
BC021277.2 | - | 5 | 852 | 4857987 |