Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303538.1 | NP_001290467.1 | 7 | 1436 | ||
NM_001303539.1 | NP_001290468.1 | 7 | 1346 | |||
NM_001303540.1 | NP_001290469.1 | 6 | 1241 | |||
NM_018273.3 | NP_060743.2 | 8 | 1541 | |||
NR_130317.1 | 6 | 1195 | ||||
GenBank mRNA | AK129801.1 | - | 8 | 1244 | ||
AK291521.1 | - | 6 | 942 | |||
AK297591.1 | - | 6 | 899 | |||
AK297610.1 | - | 7 | 1137 | |||
AK298317.1 | - | 7 | 1047 | |||
AY358161.1 | - | 6 | 933 | |||
BC016919.1 | - | 8 | 1233 | 4400601 | ||
BC040021.1 | - | 8 | 1239 |