Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FGGY SIRNA Assays ›
Gene Symbol:
FGGY
Gene Name:
FGGY carbohydrate kinase domain containing
Gene Aliases:
-
Chromosome Location:
Chr.1: 59296405 - 59762730 on Build GRCh38
UniGene ID:
Hs.444301
Species:
Human
siRNA ID
HSS124275
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FGGY
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001113411.1
NP_001106882.1
6
817
NM_001244714.1
NP_001231643.1
4
454
NM_018291.3
NP_060761.3
6
817
NR_103473.1
7
775
XM_011541730.1
XP_011540032.1
6
741
XM_011541731.1
XP_011540033.1
6
675
XM_011541732.1
XP_011540034.1
6
718
XM_011541733.1
XP_011540035.1
6
636
XM_011541735.1
XP_011540037.1
6
636
XM_011541736.2
XP_011540038.1
4
582
XM_017001643.1
XP_016857132.1
6
847
XM_017001644.1
XP_016857133.1
6
640
XM_017001645.1
XP_016857134.1
6
741
XM_017001646.1
XP_016857135.1
6
718
XM_017001647.1
XP_016857136.1
6
846
XM_017001648.1
XP_016857137.1
7
660
XM_017001649.1
XP_016857138.1
6
786
XM_017001650.1
XP_016857139.1
8
1009
XM_017001651.1
XP_016857140.1
8
865
XM_017001652.1
XP_016857141.1
6
786
XM_017001653.1
XP_016857142.1
6
718
XM_017001654.1
XP_016857143.1
4
537
XM_017001655.1
XP_016857144.1
4
446
XM_017001656.1
XP_016857145.1
7
853
XM_017001657.1
XP_016857146.1
6
641
XM_017001658.1
XP_016857147.1
7
784
XM_017001659.1
XP_016857148.1
5
674
XM_017001660.1
XP_016857149.1
5
577
XM_017001661.1
XP_016857150.1
5
734
XM_017001662.1
XP_016857151.1
5
419
XM_017001663.1
XP_016857152.1
5
594
XM_017001664.1
XP_016857153.1
4
471
XM_017001665.1
XP_016857154.1
4
378
XM_017001666.1
XP_016857155.1
4
532
XM_017001667.1
XP_016857156.1
4
586
XM_017001668.1
XP_016857157.1
4
342
XM_017001669.1
XP_016857158.1
4
437
XM_017001670.1
XP_016857159.1
4
1210
XM_017001671.1
XP_016857160.1
4
338
XM_017001672.1
XP_016857161.1
5
674
XM_017001673.1
XP_016857162.1
5
731
XM_017001674.1
XP_016857163.1
5
590
XM_017001675.1
XP_016857164.1
5
674
XM_017001676.1
XP_016857165.1
5
734
XM_017001677.1
XP_016857166.1
2
2912
XM_017001678.1
XP_016857167.1
2
4187
XM_017001679.1
XP_016857168.1
6
786
GenBank mRNA
AK022237.1
-
5
674
AK054842.1
-
4
445
AK090568.1
-
7
775
AK315649.1
-
6
702
BC014947.1
-
6
702
4871664
BX647626.1
-
6
789
Back To Top
More Information
Panther Classification:
Molecular Function -
carbohydrate kinase
Gene Ontology Categories:
Function(s)
Process(es)
carbohydrate metabolic process
phosphorylation
neuron cellular homeostasis
molecular_function
kinase activity
phosphotransferase activity, alcohol group as acceptor
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-x88h5:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline