Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCOLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253693.1 | NP_001240622.1 | 7 | 892 | ||
NM_018298.10 | NP_060768.8 | 8 | 1060 | |||
XM_005271003.1 | XP_005271060.1 | 9 | 1457 | |||
XM_006710750.1 | XP_006710813.1 | 9 | 1276 | |||
XM_011541739.2 | XP_011540041.2 | 8 | 984 | |||
XM_011541740.2 | XP_011540042.1 | 7 | 818 | |||
XM_011541741.2 | XP_011540043.1 | 8 | 991 | |||
XM_011541742.1 | XP_011540044.1 | 9 | 1194 | |||
GenBank mRNA | AF475085.1 | - | 8 | 966 | ||
AK001868.1 | - | 7 | 798 | |||
AK095148.1 | - | 2 | 935 | |||
AK292206.1 | - | 8 | 992 | |||
AK314354.1 | - | 8 | 1020 | |||
BC060765.1 | - | 9 | 1290 | 30340464 | ||
BX647497.1 | - | 10 | 1213 |