Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RHOT1 SIRNA Assays ›
Gene Symbol:
RHOT1
Gene Name:
ras homolog family member T1
Gene Aliases:
ARHT1, MIRO-1, MIRO1
Chromosome Location:
Chr.17: 32142454 - 32253374 on Build GRCh38
UniGene ID:
Hs.655325
Species:
Human
siRNA ID
HSS124304
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RHOT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001033566.2
NP_001028738.1
9, 10
888
NM_001033567.2
NP_001028739.2
10, 11
1012
NM_001033568.2
NP_001028740.1
9, 10
888
NM_001288754.1
NP_001275683.1
9, 10
888
NM_001288755.1
NP_001275684.1
9, 10
884
NM_001288758.1
NP_001275687.1
10, 11
1012
NM_018307.4
NP_060777.3
9, 10
888
NR_110083.1
9, 10
888
XM_011524969.2
XP_011523271.1
8, 9
612
XM_011524971.2
XP_011523273.2
9, 10
684
XM_011524973.2
XP_011523275.2
9, 10
684
XM_011524975.2
XP_011523277.1
10, 11
1141
XM_011524976.2
XP_011523278.2
9, 10
684
XM_017024819.1
XP_016880308.1
10, 11
916
XM_017024820.1
XP_016880309.1
8, 9
612
XM_017024821.1
XP_016880310.1
9, 10
684
XM_017024822.1
XP_016880311.1
9, 10
913
XM_017024823.1
XP_016880312.1
9, 10
684
XM_017024824.1
XP_016880313.1
10, 11
1019
XM_017024825.1
XP_016880314.1
10, 11
1111
GenBank mRNA
AJ496730.1
-
9, 10
876
AJ517412.1
-
9, 10
649
AK001902.1
-
3, 4
252
AK022695.1
-
2, 3
182
AK294407.1
-
9, 10
662
AL136929.1
-
9, 10
712
AY094972.1
-
9, 10
765
BC015698.2
-
4, 5
382
BC041114.1
-
9, 10
822
BC051818.1
-
8, 9
599
BC060781.1
-
9, 10
888
4823115
BC068463.1
-
9, 10
776
BC125104.1
-
9, 10
717
40118340
BC125105.1
-
9, 10
717
40118341
Back To Top
More Information
Panther Classification:
Molecular Function -
small GTPase
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
cellular homeostasis
mitochondrion transport along microtubule
regulation of small GTPase mediated signal transduction
mitochondrial outer membrane permeabilization
molecular_function
GTPase activity
calcium ion binding
protein binding
GTP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0