Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ACOXL SIRNA Assays ›
Gene Symbol:
ACOXL
Gene Name:
acyl-CoA oxidase-like
Gene Aliases:
-
Chromosome Location:
Chr.2: 110732539 - 111118571 on Build GRCh38
UniGene ID:
Hs.253320
Species:
Human
siRNA ID
HSS124306
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ACOXL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142807.1
NP_001136279.1
10
991
XM_011511404.2
XP_011509706.1
10
1025
XM_011511405.2
XP_011509707.1
10
1043
XM_011511406.2
XP_011509708.1
10
1016
XM_011511407.2
XP_011509709.1
9
776
XM_011511409.1
XP_011509711.1
10
1025
XM_011511411.2
XP_011509713.1
10
1025
XM_011511413.1
XP_011509715.1
10
1025
XM_011511414.2
XP_011509716.1
10
1025
XM_011511415.2
XP_011509717.1
10
1025
XM_011511416.2
XP_011509718.1
10
1025
XM_011511419.2
XP_011509721.1
10
1025
XM_011511420.2
XP_011509722.1
10
1025
XM_011511421.2
XP_011509723.1
10
1025
XM_011511422.1
XP_011509724.1
10
1025
XM_011511423.1
XP_011509725.1
10
1025
XM_011511427.2
XP_011509729.1
10
1025
XM_011511428.2
XP_011509730.1
10
1025
XM_011511429.2
XP_011509731.1
10
1025
XM_011511432.2
XP_011509734.1
10
853
XM_017004431.1
XP_016859920.1
10
1025
XM_017004432.1
XP_016859921.1
10
1025
XM_017004433.1
XP_016859922.1
2
100
XM_017004434.1
XP_016859923.1
2
60
GenBank mRNA
AK001904.1
-
4
284
AK300513.1
-
10
990
BC022268.1
-
10
930
BC131514.1
-
4
281
BX282060.1
-
2
91
Back To Top
More Information
Panther Classification:
Molecular Function -
oxidoreductase
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
fatty acid beta-oxidation using acyl-CoA dehydrogenase
fatty acid beta-oxidation using acyl-CoA oxidase
lipid homeostasis
fatty-acyl-CoA binding
molecular_function
acyl-CoA dehydrogenase activity
electron carrier activity
pristanoyl-CoA oxidase activity
flavin adenine dinucleotide binding
oxidoreductase activity, acting on the CH-CH group of donors, with a flavin as acceptor
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-phbv4:80/100.66.79.66:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline