Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPP6R3 SIRNA Assays ›
Gene Symbol:
PPP6R3
Gene Name:
protein phosphatase 6 regulatory subunit 3
Gene Aliases:
C11orf23, PP6R3, SAP190, SAPL, SAPLa, SAPS3
Chromosome Location:
Chr.11: 68460712 - 68615334 on Build GRCh38
UniGene ID:
Hs.503022
Species:
Human
siRNA ID
HSS124313
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPP6R3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164160.1
NP_001157632.1
4
509
NM_001164161.1
NP_001157633.1
4
509
NM_001164162.1
NP_001157634.1
4
509
NM_001164163.1
NP_001157635.1
4
509
NM_001164164.1
NP_001157636.1
5
614
NM_018312.4
NP_060782.2
5
614
XM_006718608.3
XP_006718671.1
5
625
XM_006718612.3
XP_006718675.1
5
625
XM_006718613.2
XP_006718676.1
5
625
XM_006718614.2
XP_006718677.1
5
625
XM_006718618.3
XP_006718681.1
5
625
XM_006718621.2
XP_006718684.1
5
625
XM_006718622.3
XP_006718685.1
5
625
XM_006718623.3
XP_006718686.1
5
608
XM_006718624.2
XP_006718687.1
5
625
XM_006718627.3
XP_006718690.1
5
625
XM_011545135.1
XP_011543437.1
5
625
XM_011545136.2
XP_011543438.1
4
508
XM_011545137.2
XP_011543439.1
5
608
XM_011545138.2
XP_011543440.1
6
826
XM_011545139.1
XP_011543441.1
5
625
XM_011545140.2
XP_011543442.1
5
625
XM_011545141.1
XP_011543443.1
5
625
XM_011545142.2
XP_011543444.1
5
625
XM_011545143.1
XP_011543445.1
5
625
XM_011545145.2
XP_011543447.1
5
625
XM_017017962.1
XP_016873451.1
4
463
XM_017017963.1
XP_016873452.1
5
549
XM_017017964.1
XP_016873453.1
5
625
XM_017017965.1
XP_016873454.1
4
508
XM_017017966.1
XP_016873455.1
5
711
XM_017017967.1
XP_016873456.1
5
625
XM_017017968.1
XP_016873457.1
4
508
XM_017017969.1
XP_016873458.1
5
625
XM_017017970.1
XP_016873459.1
4
508
XM_017017971.1
XP_016873460.1
5
625
XM_017017972.1
XP_016873461.1
4
481
XM_017017973.1
XP_016873462.1
5
608
XM_017017974.1
XP_016873463.1
5
625
XM_017017975.1
XP_016873464.1
5
625
XM_017017976.1
XP_016873465.1
4
508
XM_017017977.1
XP_016873466.1
4
396
XM_017017978.1
XP_016873467.1
3
247
XM_017017979.1
XP_016873468.1
3
247
XM_017017980.1
XP_016873469.1
3
247
XM_017017981.1
XP_016873470.1
3
247
XM_017017982.1
XP_016873471.1
3
247
XM_017017983.1
XP_016873472.1
5
628
XM_017017984.1
XP_016873473.1
4
445
GenBank mRNA
AB046778.2
-
4
457
AF264779.1
-
4
519
AF264780.1
-
4
519
AK298180.1
-
4
374
AK303315.1
-
3
248
AK315136.1
-
4
478
BC105933.1
-
3
377
40029064
BC105934.1
-
3
377
40029058
BC107599.1
-
3
377
40029065
BP314824.1
-
4
509
CN351527.1
-
5
583
CR749815.1
-
4
472
CR933658.1
-
4
472
CR936718.1
-
4
477
DQ111954.2
-
2
242
Back To Top
More Information
Panther Classification:
Molecular Function -
phosphatase modulator
Gene Ontology Categories:
Function(s)
Process(es)
regulation of phosphoprotein phosphatase activity
COPII vesicle coating
protein binding
protein phosphatase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-slhn9:80/100.66.78.111:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline