Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRIX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018321.3 | NP_060791.3 | 9, 10 | 818 | ||
GenBank mRNA | AF347667.1 | - | 9, 10 | 794 | ||
AK289610.1 | - | 9, 10 | 817 | |||
AK307492.1 | - | 5, 6 | 401 | |||
AY364244.1 | - | 9, 10 | 794 | |||
BC001546.1 | - | 9, 10 | 818 | 3462041 | ||
BC006014.1 | - | 9, 10 | 821 | |||
BC036741.1 | - | 9, 10 | 1168 | 5107954 |