Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI4K2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018323.3 | NP_060793.2 | 2 | 537 | ||
XM_005248174.1 | XP_005248231.1 | 2 | 535 | |||
XM_005248175.4 | XP_005248232.1 | 2 | 315 | |||
GenBank mRNA | AF411320.1 | - | 2 | 404 | ||
AK001967.1 | - | 2 | 525 | |||
AK128622.1 | - | 3 | 442 | |||
AY065990.1 | - | 2 | 515 | |||
AY091514.1 | - | 2 | 524 | |||
BC051749.1 | - | 2 | 537 | 6050442 |