Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTLC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018327.2 | NP_060797.2 | 2 | 520 | ||
XM_011529279.1 | XP_011527581.1 | 2 | 288 | |||
XM_017027922.1 | XP_016883411.1 | 2 | 288 | |||
GenBank mRNA | AK001974.1 | - | 2 | 300 | ||
AK300184.1 | - | 2 | 326 | |||
BC005205.1 | - | 2 | 520 | |||
BC020656.1 | - | 2 | 236 | |||
BC150644.1 | - | 2 | 479 | 9057014 | ||
CF994231.1 | - | 2 | 294 | |||
CX870585.1 | - | 1 | 13 |