Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIS18BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018353.4 | NP_060823.3 | 11 | 3038 | ||
XM_005267833.4 | XP_005267890.1 | 12 | 3085 | |||
XM_011536923.1 | XP_011535225.1 | 8 | 1523 | |||
XM_017021425.1 | XP_016876914.1 | 13 | 3066 | |||
GenBank mRNA | AB067490.1 | - | 11 | 3039 | ||
BC065544.1 | - | 11 | 2829 | 6015439 |