Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018358.2 | NP_060828.2 | 13 | 1400 | ||
XM_011512959.2 | XP_011511261.1 | 13 | 1404 | |||
GenBank mRNA | AK002060.1 | - | 13 | 1377 | ||
AK024758.1 | - | 7 | 599 | |||
AK027288.1 | - | 7 | 668 | |||
AK092415.1 | - | 13 | 1283 | |||
AK097073.1 | - | 12 | 1667 | |||
AK290106.1 | - | 13 | 1272 | |||
AK299433.1 | - | 13 | 1164 | |||
BC009253.2 | - | 13 | 1279 | 3028775 | ||
BC035652.1 | - | 13 | 1274 | |||
BC051754.2 | - | 13 | 1274 | 6064628 | ||
BC051884.1 | - | 13 | 1274 | 5105761 | ||
KU178694.1 | - | 13 | 1267 | |||
KU178695.1 | - | 13 | 1224 |