Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN7C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018362.3 | NP_060832.1 | 1, 2 | 67 | ||
GenBank mRNA | AK002077.1 | - | 1, 2 | 61 | ||
AK223156.1 | - | 1, 2 | 61 | |||
BC009774.1 | - | 1, 2 | 54 | |||
BC053907.1 | - | 1, 2 | 44 | 6502685 | ||
DA231006.1 | - | 1, 2 | 67 |