Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300953.1 | NP_001287882.1 | 9 | 716 | ||
NM_001300954.1 | NP_001287883.1 | 10 | 898 | |||
NM_001300955.1 | NP_001287884.1 | 9 | 774 | |||
NM_018367.6 | NP_060837.3 | 10 | 827 | |||
XM_011545151.2 | XP_011543453.1 | 9 | 691 | |||
XM_011545152.2 | XP_011543454.1 | 10 | 763 | |||
GenBank mRNA | AF214454.1 | - | 10 | 781 | ||
AF327353.1 | - | 10 | 792 | |||
AK002100.1 | - | 12 | 958 | |||
AK293800.1 | - | 11 | 798 | |||
AK294978.1 | - | 9 | 647 | |||
AK295142.1 | - | 10 | 815 | |||
AK315000.1 | - | 10 | 793 | |||
AK316120.1 | - | 9 | 712 | |||
BC063034.1 | - | 11 | 787 | |||
BC073853.1 | - | 10 | 744 | 3867010 |