Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252148.1 | NP_001239077.1 | 1, 2 | 808 | ||
NM_001252150.1 | NP_001239079.1 | 1, 2 | 808 | |||
NM_001252152.1 | NP_001239081.1 | 1, 2 | 82 | |||
NM_018375.4 | NP_060845.2 | 1, 2 | 808 | |||
XM_005267834.1 | XP_005267891.1 | 1, 2 | 808 | |||
XM_017021426.1 | XP_016876915.1 | 1, 2 | 808 | |||
GenBank mRNA | AK002136.1 | - | 1, 2 | 784 | ||
AK222547.1 | - | 1, 2 | 786 | |||
AK310285.1 | - | 1, 2 | 446 | |||
AK316381.1 | - | 1, 2 | 71 | |||
AY358687.1 | - | 1, 2 | 254 | |||
AY780789.1 | - | 1, 2 | 106 | |||
BC047682.1 | - | 1, 2 | 738 | 6066186 | ||
BC064383.1 | - | 1, 2 | 523 | 5737339 | ||
BP299863.1 | - | 1, 2 | 82 | |||
BX643103.1 | - | 1, 2 | 378 | |||
FJ423549.1 | - | 1, 2 | 106 |