Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252148.1 | NP_001239077.1 | 5 | 1279 | ||
NM_001252150.1 | NP_001239079.1 | 6 | 1348 | |||
NM_001252151.1 | NP_001239080.1 | 6 | 775 | |||
NM_001252152.1 | NP_001239081.1 | 7 | 763 | |||
NM_018375.4 | NP_060845.2 | 6 | 1348 | |||
XM_005267834.1 | XP_005267891.1 | 5 | 1150 | |||
XM_017021426.1 | XP_016876915.1 | 7 | 1489 | |||
GenBank mRNA | AK002136.1 | - | 6 | 1324 | ||
AK222547.1 | - | 6 | 1326 | |||
AK293222.1 | - | 6 | 768 | |||
AK316381.1 | - | 7 | 752 | |||
AY358687.1 | - | 6 | 794 | |||
AY780789.1 | - | 5 | 577 | |||
BC047682.1 | - | 6 | 1278 | 6066186 | ||
BC064383.1 | - | 6 | 1063 | 5737339 | ||
BP269907.1 | - | 5 | 550 |