Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006622.2 | NP_001006623.1 | 4 | 488 | ||
NM_001006623.2 | NP_001006624.1 | 4 | 488 | |||
NM_018383.4 | NP_060853.3 | 4 | 488 | |||
XM_005263697.2 | XP_005263754.1 | 4 | 488 | |||
XM_011511436.1 | XP_011509738.1 | 4 | 517 | |||
XM_017004435.1 | XP_016859924.1 | 4 | 488 | |||
XM_017004436.1 | XP_016859925.1 | 4 | 488 | |||
GenBank mRNA | AB044749.1 | - | 4 | 448 | ||
AK002156.1 | - | 4 | 463 | |||
AK223319.1 | - | 4 | 465 | |||
BC005401.1 | - | 4 | 472 | |||
BC013990.2 | - | 4 | 463 | |||
BC068484.1 | - | 4 | 463 | |||
BU597855.1 | - | 4 | 475 | |||
DA768238.1 | - | 4 | 488 |