Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAPTM4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018407.4 | NP_060877.3 | 6 | 985 | ||
GenBank mRNA | AF317417.1 | - | 6 | 556 | ||
AF527412.1 | - | 6 | 914 | |||
AK075326.1 | - | 6 | 747 | |||
AL136942.1 | - | 6 | 739 | |||
AY057051.2 | - | 6 | 985 | |||
AY261384.1 | - | 6 | 1004 | |||
BC014129.2 | - | 6 | 741 | 4581284 | ||
BC031021.1 | - | 6 | 766 | 4690368 | ||
BC038117.1 | - | 6 | 1533 | 5264567 |