Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEMGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018437.4 | NP_060907.2 | 4 | 657 | ||
NM_197978.2 | NP_932095.1 | 3 | 711 | |||
XM_005252086.1 | XP_005252143.1 | 4 | 619 | |||
XM_011518845.2 | XP_011517147.1 | 4 | 531 | |||
XM_011518846.2 | XP_011517148.1 | 4 | 641 | |||
GenBank mRNA | AF116617.1 | - | 1 | 113 | ||
AF130060.2 | - | 3 | 444 | |||
AF228713.2 | - | 4 | 564 | |||
AF306682.1 | - | 3 | 493 | |||
AF306683.1 | - | 3 | 459 | |||
AF322875.1 | - | 4 | 563 | |||
AK292441.1 | - | 4 | 583 | |||
AL833094.1 | - | 3 | 503 | |||
AY244805.1 | - | 3 | 657 | |||
AY255672.1 | - | 3 | 562 | |||
BC048324.1 | - | 4 | 566 | 5166612 | ||
BG571634.1 | - | 3 | 535 | |||
BG717036.1 | - | 4 | 602 |