Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1AR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128426.2 | NP_001121898.1 | 7 | 709 | ||
NM_018569.5 | NP_061039.3 | 8 | 808 | |||
NR_136622.1 | 8 | 804 | ||||
XM_017008378.1 | XP_016863867.1 | 9 | 967 | |||
XM_017008379.1 | XP_016863868.1 | 8 | 863 | |||
XM_017008380.1 | XP_016863869.1 | 8 | 491 | |||
GenBank mRNA | AK315299.1 | - | 8 | 708 | ||
AL136628.1 | - | 8 | 726 | |||
BC009485.2 | - | 7 | 615 | 3623656 | ||
BM823928.1 | - | 8 | 416 | |||
BX647702.1 | - | 8 | 761 |