Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037639.2 | NP_001032728.1 | 2 | 347 | ||
NM_001324436.1 | NP_001311365.1 | 2 | 347 | |||
NM_001324437.1 | NP_001311366.1 | 2 | 347 | |||
NM_001324438.1 | NP_001311367.1 | 2 | 347 | |||
NM_018622.6 | NP_061092.3 | 2 | 347 | |||
NR_136893.1 | 2 | 347 | ||||
XM_005247582.4 | XP_005247639.1 | 2 | 342 | |||
XM_017006800.1 | XP_016862289.1 | 2 | 342 | |||
XM_017006801.1 | XP_016862290.1 | 2 | 340 | |||
XM_017006802.1 | XP_016862291.1 | 2 | 340 | |||
XM_017006803.1 | XP_016862292.1 | 2 | 338 | |||
GenBank mRNA | AF197937.1 | - | 2 | 285 | ||
AK024765.1 | - | 2 | 319 | |||
BC003653.1 | - | 2 | 334 | 3537438 | ||
BC014058.2 | - | 2 | 305 | 3937238 | ||
BG714761.1 | - | 2 | 344 | |||
CB122278.1 | - | 2 | 344 |