Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HES6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142853.2 | NP_001136325.1 | 4 | 510 | ||
NM_001282434.1 | NP_001269363.1 | 4 | 476 | |||
NM_018645.5 | NP_061115.2 | 4 | 516 | |||
GenBank mRNA | AB035179.1 | - | 4 | 297 | ||
AF260237.1 | - | 4 | 357 | |||
AK075040.1 | - | 3 | 665 | |||
AK293111.1 | - | 4 | 430 | |||
BC007939.2 | - | 4 | 421 | 4131881 | ||
BE796220.1 | - | 4 | 516 | |||
DB495731.1 | - | 4 | 385 | |||
DT217800.1 | - | 4 | 433 |